KEY040898023 Key Organics / BIONET

4-[(1-oxo-2,3-dihydro-1H-isoindol-2-yl)methyl]benzoic acid

  • MDL number MFCD06200936
  • Cas Number : 503039-50-7
  • Molecular Weight : 267.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228 - 230
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66516

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0814_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart