KEY040898031 Key Organics / BIONET

1-(3-ethynylphenyl)-3,3-dimethylazetidin-2-one

  • MDL number MFCD06200947
  • Cas Number : 886361-64-4
  • Molecular Weight : 199.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57 - 59
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69992

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart