KEY040898034 Key Organics / BIONET

6-chloro-2-(4-hydroxyphenyl)-3-methylquinoline-4-carboxylic acid

  • MDL number MFCD06200950
  • Cas Number : 854867-53-1
  • Molecular Weight : 313.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 350
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70014

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0933_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart