KEY040898038 Key Organics / BIONET

methyl (6Z)-6-[(4-methylbenzenesulfonyl)imino]-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD06200981
  • Cas Number : 209971-44-8
  • Molecular Weight : 306.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214 - 216
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84354

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart