KEY040898039 Key Organics / BIONET

5-(4-methylbenzenesulfonyl)-2-phenylpyrimidin-4-ol

  • MDL number MFCD06200982
  • Cas Number : 885949-45-1
  • Molecular Weight : 326.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 325 - 328
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86874

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7N-585S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart