KEY040898041 Key Organics / BIONET

2-[5-(ethoxycarbonyl)-1-methyl-1H-pyrrole-3-carbonyl]benzoic acid

  • MDL number MFCD06200987
  • Cas Number : 120562-90-5
  • Molecular Weight : 301.298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91241

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8X-0729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart