KEY040898044 Key Organics / BIONET

dimethyl({6-methylimidazo[1,2-a]pyridin-3-yl}methyl)amine

  • MDL number MFCD06200990
  • Cas Number : 885949-46-2
  • Molecular Weight : 189.262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82 - 84
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91257

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8X-0813_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart