KEY040898045 Key Organics / BIONET

5-amino-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD01159705
  • Cas Number : 98589-46-9
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 274 - 276
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94207

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9X-0700_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart