KEY040898050 Key Organics / BIONET

3-(hydroxymethyl)-1,2-dihydroquinolin-2-one

  • MDL number MFCD06200995
  • Cas Number : 90097-45-3
  • Molecular Weight : 175.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97615

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/H-2032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart