KEY040898057 Key Organics / BIONET

[(4-hydroxy-3-methylphenyl)sulfanyl]formonitrile

  • MDL number MFCD00086627
  • Cas Number : 3774-53-6
  • Molecular Weight : 165.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63037

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12X-0817_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart