KEY040898092 Key Organics / BIONET

ethyl 2-(5-bromo-1H-indol-3-yl)-2-oxoacetate

  • MDL number MFCD02657946
  • Cas Number : 17826-11-8
  • Molecular Weight : 296.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248 - 249
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-5043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart