KEY040898094 Key Organics / BIONET

5-(4-methoxyphenyl)thiophene-2-carbaldehyde

  • MDL number MFCD00663639
  • Cas Number : 38401-67-1
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3513

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart