KEY040898104 Key Organics / BIONET

6-(morpholin-4-yl)pyridine-3-carboxylic acid

  • MDL number MFCD02682034
  • Cas Number : 120800-52-4
  • Molecular Weight : 208.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258 - 261
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69971

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart