KEY040898116 Key Organics / BIONET

ethyl (2E)-2-cyano-3-(pyridin-3-yl)prop-2-enoate

  • MDL number MFCD00564622
  • Cas Number : 103011-95-6
  • Molecular Weight : 202.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67 - 70
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70016

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart