KEY040898117 Key Organics / BIONET

ethyl (2E)-3-(2H-1,3-benzodioxol-5-yl)-2-cyanoprop-2-enoate

  • MDL number MFCD00022988
  • Cas Number : 2286-56-8
  • Molecular Weight : 245.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95 - 97
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70018

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0941_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart