KEY040898118 Key Organics / BIONET

ethyl (2E)-2-cyano-3-(thiophen-2-yl)prop-2-enoate

  • MDL number MFCD00039684
  • Cas Number : 31330-51-5
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79 - 82
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70020

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0943_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart