KEY040898134 Key Organics / BIONET

1-(4-acetamidobenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD02081043
  • Cas Number : 314744-44-0
  • Molecular Weight : 326.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88036

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-0049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart