KEY040898156 Key Organics / BIONET

2-oxo-1,2-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD01550054
  • Cas Number : 2003-79-4
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 330 - 332
Iupac Name 2-oxo-1,2-dihydro-3-quinolinecarboxylic acid
Inchi Code 1S/C10H7NO3/c12-9-7(10(13)14)5-6-3-1-2-4-8(6)11-9/h1-5H,(H,11,12)(H,13,14)
Inchi Key XOQQVKDBGLYPGH-UHFFFAOYSA-N

Description

Melting Point 330 - 332
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97593

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/H-2016_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart