KEY047386258 Key Organics / BIONET

2-(4-chlorophenyl)-3-nitropyridine

  • MDL number MFCD06200957
  • Cas Number : 886361-74-6
  • Molecular Weight : 234.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 -116
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73783

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-0713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart