KEY047386259 Key Organics / BIONET

2-phenylpyridine-4-carbonitrile

  • MDL number MFCD06200958
  • Cas Number : 33744-17-1
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 78 - 80
Iupac Name 2-phenylisonicotinonitrile
Inchi Code 1S/C12H8N2/c13-9-10-6-7-14-12(8-10)11-4-2-1-3-5-11/h1-8H
Inchi Key VXEVXBMHURSJRW-UHFFFAOYSA-N

Description

Melting Point 78 - 80
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73784

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-0714_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart