KEY047386264 Key Organics / BIONET

3-hydroxy-2-phenyl-1,4-dihydroquinolin-4-one

  • MDL number MFCD00451324
  • Cas Number : 31588-18-8
  • Molecular Weight : 237.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268 - 270
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76795

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0221_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart