KEY047386269 Key Organics / BIONET

2-(4-fluorophenyl)-5-nitropyridine

  • MDL number MFCD06200966
  • Cas Number : 886361-78-0
  • Molecular Weight : 218.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 136
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77415

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-0704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart