KEY047386277 Key Organics / BIONET

methyl 2-(4-formylphenoxy)-2-phenylacetate

  • MDL number MFCD06200973
  • Cas Number : 692754-44-2
  • Molecular Weight : 270.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 89
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77438

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-0816_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart