KEY047386279 Key Organics / BIONET

methyl 2-[(4-formylphenoxy)methyl]-3-nitrobenzoate

  • MDL number MFCD06200975
  • Cas Number : 885949-38-2
  • Molecular Weight : 315.281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63 - 65
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77491

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-5060_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart