KEY047386282 Key Organics / BIONET

methyl 2-(4-acetylphenoxy)-2-phenylacetate

  • MDL number MFCD06200978
  • Cas Number : 885949-39-3
  • Molecular Weight : 284.311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81107

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0801_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart