KEY047386311 Key Organics / BIONET

1,2-dimethyl-1H-indole-3-carboxylic acid

  • MDL number MFCD00756896
  • Cas Number : 20357-15-7
  • Molecular Weight : 189.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 211
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7309

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11X-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart