KEY047386322 Key Organics / BIONET

(2E)-2-cyano-3-(pyridin-3-yl)prop-2-enoic acid

  • MDL number MFCD02668152
  • Cas Number : 103029-74-9
  • Molecular Weight : 174.159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70024

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0953_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart