KEY047386323 Key Organics / BIONET

(2E)-2-cyano-3-(4-methylphenyl)prop-2-enoic acid

  • MDL number MFCD01313927
  • Cas Number : 20374-48-5
  • Molecular Weight : 187.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199 -202
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70025

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0954_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart