KEY048960493 Key Organics / BIONET

6-(4-methoxyphenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD03012770
  • Cas Number : 439096-32-9
  • Molecular Weight : 254.293

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2809

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart