KEY048960753 Key Organics / BIONET

[1,2,4]triazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD00232059
  • Cas Number : 5915-16-2
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 278 - 280
Iupac Name [1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Inchi Code 1S/C5H5N5/c6-4-1-2-7-5-8-3-9-10(4)5/h1-3H,6H2
Inchi Key QOXCEADXSTZKHA-UHFFFAOYSA-N

Description

Melting Point 278 - 280
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63341

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1D-106_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart