KEY048960792 Key Organics / BIONET

7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD03011802
  • Cas Number : 2627-59-0
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 242
Iupac Name 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Inchi Code 1S/C8H8N4O2/c1-4-2-6-10-3-5(8(13)14)7(9)12(6)11-4/h2-3H,9H2,1H3,(H,13,14)
Inchi Key FQGDCFDFDHJJSW-UHFFFAOYSA-N

Description

Melting Point 242
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65722

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0007_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart