KEY048960891 Key Organics / BIONET

4-[4-(2-hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazol-1-yl]benzoic acid

  • MDL number MFCD03848421
  • Cas Number : 861210-30-2
  • Molecular Weight : 262.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237 - 239
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0346_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart