KEY048961026 Key Organics / BIONET

4-(2-hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazole-1-carboximidamide

  • MDL number MFCD03848520
  • Cas Number : 210417-12-2
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 204 - 206
Iupac Name 4-(2-hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazole-1-carboximidamide
Inchi Code 1S/C7H12N4O2/c1-4-5(2-3-12)6(13)11(10-4)7(8)9/h10,12H,2-3H2,1H3,(H3,8,9)
Inchi Key BUUBDDAVRQDZCO-UHFFFAOYSA-N

Description

Melting Point 204 - 206
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77057

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4W-0227_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart