KEY048961144 Key Organics / BIONET

2-[1-amino-2-(3-chlorophenoxy)ethylidene]propanedinitrile

  • MDL number MFCD00139201
  • Cas Number : 343375-39-3
  • Molecular Weight : 233.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 203 - 205
Iupac Name 2-[1-amino-2-(3-chlorophenoxy)ethylidene]malononitrile
Inchi Code 1S/C11H8ClN3O/c12-9-2-1-3-10(4-9)16-7-11(15)8(5-13)6-14/h1-4H,7,15H2
Inchi Key SBWFFRVKUYJGAE-UHFFFAOYSA-N

Description

Melting Point 203 - 205
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84597

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7C-021_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart