KEY048961350 Key Organics / BIONET

4-benzyl-1,2-dihydrophthalazin-1-one

  • MDL number MFCD00139376
  • Cas Number : 32003-14-8
  • Molecular Weight : 236.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198 - 199
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84660

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7D-022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart