KEY048961414 Key Organics / BIONET

(2S)-3-amino-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD08689662
  • Cas Number : 160706-71-8
  • Molecular Weight : 129.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 122 - 124
Iupac Name (2S)-3-amino-1,1,1-trifluoro-2-propanol
Inchi Code 1S/C3H6F3NO/c4-3(5,6)2(8)1-7/h2,8H,1,7H2/t2-/m0/s1
Inchi Key RISHBQWFBUTROQ-REOHCLBHSA-N

Description

Melting Point 122 - 124
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6525

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-636_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart