KEY048961437 Key Organics / BIONET

5-(trifluoromethyl)-2,3-dihydro-1H-1,4-diazepine

  • MDL number MFCD02186431
  • Cas Number : 177545-13-0
  • Molecular Weight : 164.131

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 128
Purity 95
Storage temp. Store under N2 at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72775

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0693_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart