KEY048961679 Key Organics / BIONET

4-[3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propyl]-3-methyl-5-oxo-2,5-dihydro-1H-pyrazole-1-carboximidamide

  • MDL number MFCD05668924
  • Cas Number : 866145-62-2
  • Molecular Weight : 327.344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236 - 237
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0265_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart