KEY048961729 Key Organics / BIONET

8-methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD03407573
  • Cas Number : 874776-53-1
  • Molecular Weight : 244.173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196 - 197
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94230

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9X-0824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart