KEY048961826 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl 2,4-dichlorobenzene-1-sulfonate

  • MDL number MFCD06659543
  • Cas Number : 885949-57-5
  • Molecular Weight : 393.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79 - 81
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66477

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0271_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart