KEY048961830 Key Organics / BIONET

1,4-diacetyl-3-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD03425746
  • Cas Number : 383146-86-9
  • Molecular Weight : 182.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47 - 49
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0357_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart