KEY048961836 Key Organics / BIONET

1,4-dioxa-9-thia-12-azadispiro[4.2.4^{8}.2^{5}]tetradecane-11-carboxylic acid

  • MDL number MFCD06411612
  • Cas Number : 1030253-98-5
  • Molecular Weight : 259.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-0252_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart