KEY048961842 Key Organics / BIONET

6-[(4-chlorophenyl)methyl]-5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD06411616
  • Cas Number : 885949-37-1
  • Molecular Weight : 287.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 300
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77447

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart