KEY048961851 Key Organics / BIONET

4-{4-[(2E)-3-phenylprop-2-en-1-yl]piperazine-1-carbonyl}aniline

  • MDL number MFCD06659559
  • Cas Number : 219989-07-8
  • Molecular Weight : 321.424

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81096

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0223_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart