KEY048961854 Key Organics / BIONET

methyl 4-(5-aminopyridin-2-yl)benzoate

  • MDL number MFCD06411623
  • Cas Number : 223127-54-6
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 189 - 192
Iupac Name methyl 4-(5-amino-2-pyridinyl)benzoate
Inchi Code 1S/C13H12N2O2/c1-17-13(16)10-4-2-9(3-5-10)12-7-6-11(14)8-15-12/h2-8H,14H2,1H3
Inchi Key CUVFODCVMGKLTH-UHFFFAOYSA-N

Description

Melting Point 189 - 192
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81102

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5Y-0702_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart