KEY048961858 Key Organics / BIONET

ethyl 2-methyl-5-oxo-1-[2-(thiophen-2-yl)ethyl]-4,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD06659564
  • Cas Number : 885949-74-6
  • Molecular Weight : 279.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81114

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0816_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart