KEY048961861 Key Organics / BIONET

1-{2-hydroxy-4-[(4-methylphenyl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD06659565
  • Cas Number : 885949-75-7
  • Molecular Weight : 256.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81121

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart