KEY048961865 Key Organics / BIONET

tert-butyl 4-{[4-(propan-2-yl)phenyl]amino}piperidine-1-carboxylate

  • MDL number MFCD06659568
  • Cas Number : 241499-44-5
  • Molecular Weight : 318.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 99 -100
Iupac Name tert-butyl 4-(4-isopropylanilino)-1-piperidinecarboxylate
Inchi Code 1S/C19H30N2O2/c1-14(2)15-6-8-16(9-7-15)20-17-10-12-21(13-11-17)18(22)23-19(3,4)5/h6-9,14,17,20H,10-13H2,1-5H3
Inchi Key ZQGATDGLYABDTH-UHFFFAOYSA-N

Description

Melting Point 99 -100
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84451

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Y-0211_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart