KEY048961869 Key Organics / BIONET

3-(2H-1,3-benzodioxol-5-yl)-5-methyl-1H-pyrazole

  • MDL number MFCD06411629
  • Cas Number : 937022-12-3
  • Molecular Weight : 202.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141 - 143
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84464

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Y-0807_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart