KEY048961874 Key Organics / BIONET

1-{3-fluoro-4-[(4-nitrophenyl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD06659573
  • Cas Number : 885949-79-1
  • Molecular Weight : 289.262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 150
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Y-0922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart